Cole-Parmer Fluorescent Flt ORange Dye Concentrate, Pint
Representative image only.
Cole-Parmer Fluorescent Flt ORange Dye Concentrate, Pint
Cole-Parmer | Item: 0029819
Determine water flow in closed circuits or open channels
$76.91CAD / Pint of 1
$76.91CAD / Pint of 1
Product Description
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more. Orange (xanthene) dye appears as bright yellow under ultraviolet light. Minimum detectable concentration of all dyes in clear water is 1 ppm.
Tracers are available in tablets, liquid, or powder form. Quick-dissolving tablets are easy to handle with less mess than other forms. They are perfect for applications such as storm lines, cooling and plumbing systems, and sewer lines. Choose liquid for applications such as metered input, flow/dilution/disperation rate assessment, septic inspections, or fluorometric dye tests. Powdered form is well-suited for large applications where liquid is undes
NSF international certified
Specifications
  • Reagent PlatformLiquid concentrate
  • Gallons Of Water For 1 ppm (Gal)4000
  • DescriptionFluorescent Flt ORange Dye Concentrate, Pint