Cole-Parmer Fixed-Volume Pipettor, 10 µL
Representative image only.
Cole-Parmer Fixed-Volume Pipettor, 10 µL
Cole-Parmer | Item: 1597069
Ergonomic finger grip for comfortable use
$60.41CAD / Each
$60.41CAD / Each
Buy It Together
Current Item
Cole-Parmer Fixed-Volume Pipettor, 10 µL
Cole-Parmer Fixed-Volume Pipettor, 10 µL
1597069
$60.41CAD / Each
+ + + +
Cole-Parmer Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
2450223
$55.69CAD / Bag of 1000
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
$66.60CAD / Bag of 1000
+ =
Total Price 1994.08 ADD 7 ITEMS TO CART
Product Description
Includes
Color-coding drop and removal tool, calibration tool and silicon grease, along with sample tips and calibration report.
Key Features
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • 3 year warranty
More About this Item
These pipettors are offered in a wide-range of fixed volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
3 Year Warranty
Specifications
  • AutoclavableYes
  • Max Volume (µL)10
  • Accuracy (%)±0.8
  • Precision (%)±0.80
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYes
  • DescriptionFixed-Volume Single Channel Pipettor