Cole-Parmer FHD-400 Advanced Ductless Fume Hood, 59" Width; 115 VAC, 60 Hz

Representative image only.

Cole-Parmer FHD-400 Advanced Ductless Fume Hood, 59" Width; 115 VAC, 60 Hz

Cole-Parmer | Item: 7890224

Advanced carbon filtration technology with filter saturation sensor offers environmentally safe alternative to conventional ducted fume hoods

$12,892.16CAD / Each

$12,892.16CAD / Each

Buy It Together

Current Item

Cole-Parmer FHD-400 Advanced Ductless Fume Hood, 59" Width; 115 VAC, 60 Hz
Cole-Parmer FHD-400 Advanced Ductless Fume Hood, 59" Width; 115 VAC, 60 Hz
7890224

$12,892.16CAD / Each
+ +
Cole-Parmer Airflow Meter Vaneometer
7890012

$167.23CAD / Each
+ + + =
Total Price 27549.07 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Traps chemical vapors to prevent ecological impact through release into the environment
  • Cabinet airflow and face velocity offer high operator protection from fume and particle hazards
  • Ductless fume hoods are self-contained and do not require venting to the outside
  • Low airflow alarm indicates when filters need to be changed
  • Improved clamping eliminates bypass leakage
  • 3 year warranty

More About this Item

The FHD-400 Advanced Series ductless fume hoods are a series of high-efficiency products designed to protect the user and the environment from hazardous vapors generated on the work surface. At the heart of the fume hood product line is an innovative filtration technology that creates a safe work environment over the widest range of applications in the industry. These hoods contain a pre-filter, main activated carbon filter, and safety activated carbon filter. The system permits a customized combination and configuration of additional filter media (sold separately, see accessories below) for chemical and physical adsorption specific to each application.

The cabinet frame and work surfaces are durable and chemically resistant. A control panel displays the airflow and uses an electronic gas sensor to detect when the filter needs changed. Audible and visual alarms alert you to filter saturation and if the airflow reaches preset thresholds. The fume hoods comply with current technical and safety regulations.

Conformite EuropeenneCanadian Standards Association3 Year Warranty
Specifications
  • Exterior Width (in)59
  • Exterior Height (in)53
  • Exterior Depth (in)27.375
  • Interior Height (in)38
  • Power (VAC)115
  • Power (Hz)60
  • Shipping Weight (lbs)350
  • Shipping Weight (kg)159
  • Filter TypeCarbon
  • DescriptionDuctless Fume Hood, 59" Width; 115 VAC, 60 Hz
  • Warranty3 Years
Reviews
Q&A