Cole-Parmer EX Adjustable Volume Pipette, 100-1000 uL; Each

Representative image only.

Cole-Parmer EX Adjustable Volume Pipette, 100-1000 uL; Each

Cole-Parmer | Item: 0790911

Autoclavability and enhanced UV resistance provides added protection from biohazardous elements

$752.90CAD / Each

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$752.90CAD / Each

Buy It Together

Current Item

Cole-Parmer EX Adjustable Volume Pipette, 100-1000 uL; Each
Cole-Parmer EX Adjustable Volume Pipette, 100-1000 uL; Each
0790911

$752.90CAD / Each
+ +
Cole-Parmer Pipette Tips, 1-10 mL, White; 200/Pk
0790925

$82.95CAD / Bag of 200
+ + + + +
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
2450243

$58.05CAD / Bag of 1000
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261

$66.60CAD / Bag of 1000
=
Total Price 2516.55 ADD 9 ITEMS TO CART
Product Description

Includes

calibration document supplied by the manufacturer. Order tips separately.

Key Features

  • Locking lever prevents accidental volume change
  • Self-lubricating PTFE seal-ring eliminates routine lubrication
  • Sterilize by autoclave or 254 nm UV light
  • 2 year warranty

More About this Item

This lightweight, slender pipettor features molded channels to improve grip and its smooth plunger action reduces operator fatigue. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.

Two year warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)1000
  • Min Volume (µL)100
  • Min Volume (mL)0.1
  • Max Volume (mL)1
  • Min Dispensing (mL)0.1
  • Max Dispensing (mL)1
  • Increments (µL)1
  • Accuracy (%)±1.00% to ±0.70%
  • Number Of Channels1
  • Tip EjectorYes
  • Body MaterialDS polyester urethane zzle filter; Al2O3ceramic and PP plunger assembly
  • DescriptionAdjustable Volume Pipette, 100-1000 uL; Each
  • Warranty2 year
Reviews
Q&A