Cole-Parmer Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
Representative image only.
Cole-Parmer Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
Cole-Parmer | Item: 2869050
Indivdual needles provide equal flow of gas for faster, consistent evaporation of samples
$2,762.61CAD / Each
$2,762.61CAD / Each
Buy It Together
Current Item
Cole-Parmer Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
Cole-Parmer Evaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
2869050
$2,762.61CAD / Each
+ + + + + =
Total Price 28692.80 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Economical and reliable evaporation in a compact instrument
  • Models available to accommodate different-sized tubes and microwell plates
  • 3 year warranty
More About this Item
Rapidly evaporate common organic solvents—ideal for pharmaceutical, environmental, HPLC, GCMS, and general research applications. Needle valve controls the flow of gas into the equalizer manifold, which provides equal gas flow and uniform evaporation to each well. Needle valves ensure control for fast concentration of multiple samples. Each needle is made from 15-gauge stainless steel with easy height adjustment to accommodate a variety of sample sizes.
Stand-Alone Evaporator/Concentrators are made of autoclavable polypropylene with a nickel-plated valve; feature cutouts for easy removal of plates. Select either a 96- or 384-well model.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
3 Year Warranty
Specifications
  • Condenser TypeEvaporator
  • Total Positions96
  • DescriptionEvaporator/Concentrator for 96-well Plates; stand-alone (non-heated)
  • Warranty3 Years