Cole-Parmer Elite PAP Pen - Broad Point
Representative image only.
Cole-Parmer Elite PAP Pen - Broad Point
Cole-Parmer | Item: 7595553
Keep specimens in place
$145.72CAD / Each
$145.72CAD / Each
Product Description
Key Features
  • Resist acetone and alcohol once dry
  • Highly visible red-tint color
  • Clean off with xylene and xylene substitutes
More About this Item
Draw your own hydrophobic well on slides to prevent reagent waste. The liquid-repellent barrier creates surface tension, keeping critical specimens in place.
Each pen contains 13 mL of fluid—good for hundreds of wells!
Specifications
  • ColorRed
  • DescriptionHydrophobic Barrier PAP Pen; 1/Ea