Cole-Parmer Electronic Peltier Cuvette Holder
Representative image only.
Cole-Parmer Electronic Peltier Cuvette Holder
Cole-Parmer | Item: 8307035
Optimize performance of your Cole-Parmer® spectrophotometer
$2,637.37CAD / Each
$2,637.37CAD / Each
Buy It Together
Current Item
Cole-Parmer Electronic Peltier Cuvette Holder
Cole-Parmer Electronic Peltier Cuvette Holder
8307035
$2,637.37CAD / Each
+ + +
Cole-Parmer SP-200-VIS Visible Spectrophotometer; 110 VAC, 60 Hz
7900064
$4,508.58CAD / Each
$5,635.72CAD / Each
+ + + +
Cole-Parmer SP-300-VIS Visible Spectrophotometer; 90 to 264 VAC
8305815
$6,447.56CAD / Each
$8,059.44CAD / Each
+ + =
Total Price 67291.24 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Expand analysis capabilities or make testing easier
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
The single position electronic Peltier cell holder is compatible with the 73 series, as well as the Genova Plus and Nano lines of spectrophotometers.
Specifications
  • Compatible WithCole-Parmer SP-300, SP-350 Spectrophometers
  • DescriptionElectronic Peltier Cuvette Holder