Cole-Parmer EC91346S 4-electrode conductivity cell, 3/4" NPT(M)
Representative image only.
Cole-Parmer EC91346S 4-electrode conductivity cell, 3/4" NPT(M)
Cole-Parmer | Item: 1950300
Four-cell electrode eliminates polarizing effects.
$770.88CAD / Each
BUY MORE & SAVE
  • 1-5$770.88 CAD / Each
  • 6-11$693.79 CAD / Each
  • 12-23$655.25 CAD / Each
  • 24+$616.70 CAD / Each
$770.88CAD / Each
BUY MORE & SAVE
  • 1-5$770.88 CAD / Each
  • 6-11$693.79 CAD / Each
  • 12-23$655.25 CAD / Each
  • 24+$616.70 CAD / Each
Product Description
Key Features
  • Ensures more accurate measurements in highly conductive samples
  • Measurement range of 0.5 uS to 200 uS/cm
  • Integral 100 ΩPt RTD temperature sensor
  • 1 year warranty
More About this Item
The four-cell electrode is made of epoxy with 416 SS contacting electrodes. 3/4" fittings at both ends allow for insertion into a T-fitting or submersion installations. Withstands pressures up to 6 bar (90 psi).
1 Year Warranty
Specifications
  • Cell ConstantK = 0.3
  • Cell TypeContact
  • Electrode Material416 SS
  • Body MaterialEpoxy
  • Fitting¾" NPT(M)
  • Mounting TypeThreaded
  • Min temperature (° C)-5
  • Max temperature (° C)80
  • Min temperature (° F)23
  • Max temperature (° F)176
  • Cable Length (m)10
  • Cable Length (ft)30
  • Description4-electrode conductivity cell, 3/4" NPT(M)