Cole-Parmer Disposable Dissecting Scalpels, #21 Blade; 10/Pack
Representative image only.
Cole-Parmer Disposable Dissecting Scalpels, #21 Blade; 10/Pack
Cole-Parmer | Item: 1081841
$60.70CAD / Pkg of 10
$60.70CAD / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Disposable Dissecting Scalpels, #21 Blade; 10/Pack
Cole-Parmer Disposable Dissecting Scalpels, #21 Blade; 10/Pack
1081841
$60.70CAD / Pkg of 10
+ =
Total Price 180.04 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Superior dissecting with stainless steel razor-sharp blades. These single-use scalpels are individually packaged in sealed pouches. Each scalpel includes a blade safety sleeve to protect you and the blade.
Note: Not intended for medical applications or patient use.
Specifications
  • Blade Type21
  • Grade TypeDissecting
  • MaterialStainless Steel
  • DescriptionDisposable Dissecting Scalpels, #21 Blade; 10/Pack