Cole-Parmer Disposable Dissecting Safety Scalpels, #10 Blade; 10/Pack
Representative image only.
Cole-Parmer Disposable Dissecting Safety Scalpels, #10 Blade; 10/Pack
Cole-Parmer | Item: 1081890
$143.36CAD / Pkg of 10
$143.36CAD / Pkg of 10
Product Description
More About this Item
These single-use scalpels come with fully retractable blades. The blades lock securely into place — eliminating accidents.
Note: Not intended for medical applications or patient use.
Specifications
  • Blade Type10
  • Grade TypeDissecting
  • MaterialStainless Steel
  • DescriptionDisposable Dissecting Safety Scalpels, #10 Blade; 10/Pack