Cole-Parmer Diol HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA

Representative image only.

Cole-Parmer Diol HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA

Cole-Parmer | Item: 1647171

Designed to aid in the separation and retention of very polar analytes

$975.38CAD / Each

$975.38CAD / Each

Buy It Together

Current Item

Cole-Parmer DIOL HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA
Cole-Parmer DIOL HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA
1647171

$975.38CAD / Each
+
Cole-Parmer HPLC In-Line Filters, 2 µm; 10/Pk
1647091

$2,055.38CAD / Pkg of 10
=
Total Price 3030.76 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Use to increase sensitivity in MS analysis
  • Provides alternate selectivity to that achieved with reversed phase chemistries
  • Can be used for HILIC or normal phase mode
  • Reduced extraction and dry down times
  • 3 µm particle size
  • Measures 50 mm L x 2.1 mm ID
  • 1 year warranty

More About this Item

These hydrophilic chromatographic columns have diol groups bonded to a silica matrix to separate polar to highly polar compounds that have poor retention on reversed-phase columns. Obtain good retention with eluents containing high concentrations of organic solvents. It has a good separation effect on polar compounds, and it is also used for separation of peptides, proteins, and polar drug molecules.

Columns come in 316L stainless steel (SS) tubing hardware. A SS female end fitting with a SS and PEEK frit system keeps the stationary phase in place and packed bed from disturbance. The SS end fitting accepts a male 10-32 threaded nut with standard 1/16" OD tubing.

Use the guard cartridges to maintain chromatographic integrity. They ensure erroneous materials are kept out of the more important and expensive analytical columns, leading to more accurate results. Simply match the sizes to select the correct guard cartridge for your column; guard cartridge holder 16470-89 is required to use the guard cartridges. In-line filters are available for your UPHLC and HPLC columns—order separately.

1 Year Warranty
Specifications
  • PhaseDiol
  • Carbon Loading4%
  • Column Length (mm)50
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)3
  • Max Pressure (PSI)5800
  • Max Operating Temperature (° C)60
  • DescriptionDiol HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA
  • Warranty1 Year
Reviews
Q&A