Cole-Parmer Diamond Grip Nitrile Gloves, Size 9, Double-Layer; 12 Pair/Bag
Representative image only.
Cole-Parmer Diamond Grip Nitrile Gloves, Size 9, Double-Layer; 12 Pair/Bag
Cole-Parmer | Item: 4021613
Offers superior wet and dry grip
$69.54CAD / Pkg of 12
BUY MORE & SAVE
  • 1-11$69.54 CAD / Pkg of 12
  • 12+$62.59 CAD / Pkg of 12
$69.54CAD / Pkg of 12
BUY MORE & SAVE
  • 1-11$69.54 CAD / Pkg of 12
  • 12+$62.59 CAD / Pkg of 12
Product Description
Key Features
  • Hypoallergenic
  • USDA compliant
More About this Item
Embossed diamond pattern provides superior wet and dry grip. Latex-free nitrile gloves resist punctures, cuts, snags, and abrasion. Silicone free—suitable for use in paint and powder coat applications. Hypoallergenic. Compliance: USDA accepted per 21CFR 177.2600 for food and drug handling, manufactured under ISO 9002 standards.
Specifications
  • MaterialNitrile
  • Thickness (mil)15
  • Size9
  • Length (in)12
  • Length (cm)30.5
  • ColorGreen
  • ComplianceFDA 21 CFR 177.2600; ISO 9002
  • SterileNo
  • DescriptionDiamond Grip Nitrile Gloves, Size 9, Double-Layer; 12 Pair/Bg