Cole-Parmer Polyvinyl Chloride (PVC) Tubing 3/8" x 5/8", 50 ft 

Representative image only.

Cole-Parmer Polyvinyl Chloride (PVC) Tubing 3/8" x 5/8", 50 ft 

Cole-Parmer | Item: 5014305

Specifically designed for therapeutic applications such as cell culture and gene therapy

$180.86CAD / Pkg of 1

$180.86CAD / Pkg of 1
Product Description

Key Features

  • Compatible with aseptic welding and sealing machines
  • DEHP, phthalates, and BPA free, and animal-derived ingredient free (ADIF)
  • Crystal clear tubing for easy process monitoring
  • Sterilize by gamma irradiation (<45 kGy), EtO, and E-beam
  • USP Class VI, ISO 10993 Sections 4 and 5), REACH, and RoHS compliant

More About this Item

Specifically designed for cell and gene therapy processes, Cole-Parmer DEHP-free PVC tubing provides unmatched reliability and quality. Superior toxicological and physiochemical profile, gamma stability up to 45 kGy, and compatibility with aseptic welders or sealers, make the tubing perfect for pharmaceutical and bioprocessing environments.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Specifications
  • Tubing FormulationPVC
  • Tubing OD (in)5/8
  • Tubing ID (in)3/8
  • Wall Thickness (in)1/8
  • Clarity (clear)Clear
  • Working Pressure (PSI)40
  • Working Pressure (bar)2.8
  • Max Vacuum (in Hg)29.5
  • Max Vacuum (mm Hg)749.3
  • Bend Radius (in)1 1/8
  • Bend Radius (mm)28.6
  • Length (ft)50
  • Sterilization MethodE-Beam;EtO;Gamma
  • Regulatory ApprovalsNon-Phthalate;RoHS;USP;Non-DEHP
  • Durometer65 Shore A
  • DescriptionDEHP-Free, Gamma-Stable PVC Tubing, 3/8" ID x 5/8" OD; 50 Ft 
Reviews
Q&A