Cole-Parmer Crucible Cover, High Alumina, 100 mL
Representative image only.
Cole-Parmer Crucible Cover, High Alumina, 100 mL
Cole-Parmer | Item: 6310166
Protect your samples
$16.06CAD / Each
BUY MORE & SAVE
  • 1-5$16.06 CAD / Each
  • 6-29$14.45 CAD / Each
  • 30-119$13.65 CAD / Each
  • 120+$12.85 CAD / Each
$16.06CAD / Each
BUY MORE & SAVE
  • 1-5$16.06 CAD / Each
  • 6-29$14.45 CAD / Each
  • 30-119$13.65 CAD / Each
  • 120+$12.85 CAD / Each
Buy It Together
Current Item
Cole-Parmer Crucible Cover, High Alumina, 100 mL
Cole-Parmer Crucible Cover, High Alumina, 100 mL
6310166
$16.06CAD / Each
+ =
Total Price 103.73 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of high alumina for enhanced chemical resistance Autoclavable
  • 1 year warranty
More About this Item
Compatible with the Cole-Parmer® High Alumina Tall-Form Crucible, models 63101-00 through -08. Crucible sold separately.
1 Year Warranty
Specifications
  • DescriptionCrucible Cover, High Alumina, 100 mL, 1/Ea