Cole-Parmer Class 1 Ductless Workstation, 18" Deep, 115V

Representative image only.

Cole-Parmer Class 1 Ductless Workstation, 18" Deep, 115V

Cole-Parmer | Item: 3373022

Portable, compact workstations are completely self-contained

$2,703.67CAD / Each

$2,703.67CAD / Each

Buy It Together

Current Item

Cole-Parmer Class 1 Ductless Workstation, 18" Deep, 115V
Cole-Parmer Class 1 Ductless Workstation, 18" Deep, 115V
3373022

$2,703.67CAD / Each
+ =
Total Price 3293.03 ADD 2 ITEMS TO CART
Product Description

Includes

Prefilter and carbon filter.

Key Features

  • Integral recessed work surface contains spills
  • Top airflow is 100 CFM and has high, medium and low settings
  • 1 year warranty

More About this Item

Compact benchtop workstations are ideal for your general chemistry, tissue research, forensic applications, and academic demonstrations. The prefilter and activated carbon filter remove fumes, odors, and nonhazardous chemicals.

Clear acrylic enclosure lets you easily view workstation contents from three sides. Front cutaways allow ergonomic use.

Larger workstations 33730-26 and -28 have a hinged sash that enables you to have a completely open front.

Filters for both size workstations are easily replaced by removing the prefilter and then unclipping the carbon filter from the rear baffle. Workstations 33730-26 and -28 feature a filter change light, but it is recommended filters be changed every 60 to 90 days with normal use. Models 33730-22 and -24 have an electrical port exit on the left side. Models 33730-26 and -28 have electrical port exits on both sides of workstation.

UL Listed.1 Year Warranty
Specifications
  • Hood Width (in)12
  • Exterior Width (in)12
  • Exterior Height (in)14.5
  • Exterior Depth (in)18
  • Interior Width (in)11
  • Power (VAC)115
  • Filter TypeCarbon and particulate pre-filter
  • DescriptionClass 1 Ductless Workstation, 18" Deep, 115V
  • Warranty1 Year
Reviews
Q&A