Cole-Parmer Blue Polyurethane Bench Height Ergonomic Chair With Arm Rests, Foot Ring, And Casters
Representative image only.
Cole-Parmer Blue Polyurethane Bench Height Ergonomic Chair With Arm Rests, Foot Ring, And Casters
Cole-Parmer | Item: 9000485
Ergonomic design to improve productivity
$931.18CAD / Each
BUY MORE & SAVE
  • 1-3$931.18 CAD / Each
  • 4-9$838.06 CAD / Each
  • 10+$791.50 CAD / Each
$931.18CAD / Each
BUY MORE & SAVE
  • 1-3$931.18 CAD / Each
  • 4-9$838.06 CAD / Each
  • 10+$791.50 CAD / Each
Buy It Together
Current Item
Cole-Parmer Blue Polyurethane Bench Height Ergonomic Chair with arm rests, foot ring, and casters
Cole-Parmer Blue Polyurethane Bench Height Ergonomic Chair with arm rests, foot ring, and casters
9000485
$931.18CAD / Each
+ =
Total Price 1057.45 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Chairs are available with or without arm rests
  • The six-way adjustable arm rests fit all possible ergonomic seating configurations
  • Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • Rugged polyurethane is self-skinning, resistant of chemicals, punctures and soil
  • Easily cleaned
  • Blue or black chairs with black, non-marring bell glides suitable for hard and carpeted floors
  • Warranty 15-years parts, lifetime pneumatic
More About this Item
Polyurethane chairs have a seat tilt of eight degrees forward and three degrees backward. The seat tilt locks into place. With a rugged curved back and tilting, and articulating seat, the chair offers excellent support and maximum comfort
Specifications
  • Dimension20-27.5" adjustable height (measured from the top of the seat to the floor)
  • MaterialPolyurethane
  • ColorBlue
  • DescriptionBlue Polyurethane Bench Height Ergonomic Chair with arm rests, foot ring, and casters
  • WarrantyWarranty 15-years parts, 5-years upholstery, lifetime pneumatic