Cole-Parmer BIO C4 HPLC Column, 5 µm, 300 Å, 150 mm L x 4.6 mm ID; 1/EA

Representative image only.

Cole-Parmer BIO C4 HPLC Column, 5 µm, 300 Å, 150 mm L x 4.6 mm ID; 1/EA

Cole-Parmer | Item: 1647077

C4 columns with optimized pore size provide robust separation of larger proteins and peptides

$994.54CAD / Each

$994.54CAD / Each

Buy It Together

Current Item

Cole-Parmer BIO C4 HPLC Column, 5 µm, 300 Å, 150 mm L x 4.6 mm ID; 1/EA
Cole-Parmer BIO C4 HPLC Column, 5 µm, 300 Å, 150 mm L x 4.6 mm ID; 1/EA
1647077

$994.54CAD / Each
+
Cole-Parmer HPLC In-Line Filters, 2 µm; 10/Pk
1647091

$2,055.38CAD / Pkg of 10
=
Total Price 3049.92 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 300 Å pore size is optimized for peptides and proteins
  • Use for analyzing large hydrophobic molecules
  • Provides a high density bonding for a chemically stable, robust, and low bleed phase
  • Fully scalable
  • 5 µm particle size
  • Measures 150 mm L x 4.6 mm ID
  • 1 year warranty

More About this Item

The importance of pore size is critical in the separation of large peptides and proteins. The use of a typical analytical silica template can lead to poor resolution, sensitivity, and recovery of biomolecules. The BIO C4 columns, specifically designed for large peptides and proteins sharp peak shapes, excellent analyte recovery, great reproducibility, and high sensitivity. The optimized bonding process of the BIO C4 ligand increases the surface coverage and removes secondary interactions leading to improved lifetime over other C4 columns.

Columns come in 316L stainless steel (SS) tubing hardware. A SS female end fitting with a SS and PEEK frit system keeps the stationary phase in place and packed bed from disturbance. The SS end fitting accepts a male 10-32 threaded nut with standard 1/16" OD tubing. Order in-line filters to protect your BIO C4 HPLC and UHPLC columns.

1 Year Warranty
Specifications
  • PhaseBIO C4
  • Carbon Loading5%
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • Max Pressure (PSI)8940
  • Max Operating Temperature (° C)60
  • DescriptionBIO C4 HPLC Column, 5 µm, 300 Å, 150 mm L x 4.6 mm ID; 1/EA
  • Warranty1 Year
Reviews
Q&A