Cole-Parmer BIO C18 UHPLC Column, 1.7 µm, 300 Å, 50 mm L x 2.1 mm ID; 1/EA

Representative image only.

Cole-Parmer BIO C18 UHPLC Column, 1.7 µm, 300 Å, 50 mm L x 2.1 mm ID; 1/EA

Cole-Parmer | Item: 1647075

C18 columns with optimized pore size provide robust separation of proteins and peptides

$1,049.05CAD / Each

$1,049.05CAD / Each

Buy It Together

Current Item

Cole-Parmer BIO C18 UHPLC Column, 1.7 µm, 300 Å, 50 mm L x 2.1 mm ID; 1/EA
Cole-Parmer BIO C18 UHPLC Column, 1.7 µm, 300 Å, 50 mm L x 2.1 mm ID; 1/EA
1647075

$1,049.05CAD / Each
+
Cole-Parmer UHPLC In-Line Filters, 0.5 µm; 4/Pk
1647093

$1,062.31CAD / Pkg of 4
=
Total Price 2111.36 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 300 Å pore size is optimized for peptides and proteins
  • Use for separating hydrophilic proteins
  • Based on traditional silica and bonded technology
  • Fully scalable
  • Operates up to 18,000 psi for increased efficiency at high linear velocities
  • 1.7 µm particle size
  • Measures 100 mm L x 2.1 mm ID
  • 1 year warranty

More About this Item

The importance of pore size is critical in the separation of large peptides and proteins. The use of a typical analytical silica template can lead to poor resolution, sensitivity, and recovery of biomolecules. The BIO C18 columns, specifically designed for large peptides and proteins provide sharp peak shapes, excellent analyte recovery, great reproducibility, and high sensitivity. The high surface area reduces any secondary negative interactions which could compromise peak shapes.

Columns come in 316L stainless steel (SS) tubing hardware. A SS female end fitting with a SS and PEEK frit system keeps the stationary phase in place and packed bed from disturbance. The SS end fitting accepts a male 10-32 threaded nut with standard 1/16" OD tubing. Order in-line filters to protect your BIO C18 HPLC and UHPLC columns.

1 Year Warranty
Specifications
  • PhaseBIO C18
  • Carbon Loading11%
  • Column Length (mm)50
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)1.7
  • Max Pressure (PSI)15000
  • Max Operating Temperature (° C)60
  • DescriptionBIO C18 UHPLC Column, 1.7 µm, 300 Å, 50 mm L x 2.1 mm ID; 1/EA
  • Warranty1 Year
Reviews
Q&A