Cole-Parmer Bath Thermal Transfer Fluid, Ethylene Glycol / DI Water Blend – 50:50 Mix; 4L Poly Bottle
Representative image only.
Cole-Parmer Bath Thermal Transfer Fluid, Ethylene Glycol / DI Water Blend – 50:50 Mix; 4L Poly Bottle
Cole-Parmer | Item: 7893017
$138.20CAD / Each
BUY MORE & SAVE
  • 1-3$138.20 CAD / Each
  • 4+$124.38 CAD / Each
$138.20CAD / Each
BUY MORE & SAVE
  • 1-3$138.20 CAD / Each
  • 4+$124.38 CAD / Each
Product Description
Key Features
  • 50/50 Ethylene Glycol/Water
  • Thermal transfer fluid for -25C to 100C
  • For use with circulating baths and chillers
  • 10 year warranty
More About this Item
Use bath fluids to maintain desired bath temperatures outside the range of water.
Lower freezing point than water.
10 Year Warranty
Specifications
  • Max Temperature (° C)100
  • Min Temperature (° C)-25
  • DescriptionBath Thermal Transfer Fluid, Ethylene Glycol / DI Water Blend – 50:50 Mix; 4L Poly Bottle
  • Warranty10 year