Cole-Parmer Automatic Turet, 8 Cell
Representative image only.
Cole-Parmer Automatic Turet, 8 Cell
Cole-Parmer | Item: 8307031
Optimize performance of your Cole-Parmer® spectrophotometer
$3,860.28CAD / Each
$3,860.28CAD / Each
Buy It Together
Current Item
Product Description
Key Features
  • Expand analysis capabilities or make testing easier
  • Accepts 10 x 10 pathlength cuvettes
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
The multiposition cell/cuvette holder is compatible with the 73 series, as well as the Genova Plus and Nano lines of spectrophotometers.
Specifications
  • Compatible WithCole-Parmer SP-300, SP-350 Spectrophometers
  • DescriptionAutomatic Turet, 8 Cell