Cole-Parmer Autoclavable Biohazard Bags, Red, 8.5" x 11", PP; 100/PK

Representative image only.

Cole-Parmer Autoclavable Biohazard Bags, Red, 8.5" x 11", PP; 100/PK

Cole-Parmer | Item: 1400307

Heavy-gauge, 2.25 mil thick polypropylene (PP) bags offer excellent puncture resistance

$54.52CAD / Pkg of 100

$54.52CAD / Pkg of 100

Buy It Together

Current Item

Cole-Parmer Autoclavable Biohazard Bags, Red, 8.5" x 11", PP; 100/PK
Cole-Parmer Autoclavable Biohazard Bags, Red, 8.5" x 11", PP; 100/PK
1400307

$54.52CAD / Pkg of 100
+
MTC Bio BenchBin™ Benchtop Biohazard Bin
1400304

$66.45CAD / Each
=
Total Price 120.97 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Withstand temperatures up to 136°C (277°F)
  • Prominently marked with the words “Hazardous Waste” and the “Biohazard” symbol
  • White marking area for quick and easy labeling
  • 0.43 gallon capacity
  • 8.5"W x 11"L (21.6 x 28 cm)
  • Pack of 100 bags

More About this Item

These high-temperature autoclavable bags are extremely pliable yet offer exceptional strength and durability. Seamless bottoms ensure leak-free autoclaving. Opaque "biohazard" red color along with the words "Hazardous Waste" and the "biohazard" symbol conforms to OSHA standard (29CFR1910.1030). An oversized white marking area makes it easy to label each bag with a permanent marker.

Specifications
  • Capacity (Gal)0.43
  • MaterialPolypropylene
  • ColorRed
  • Length (in)11
  • Width (in)8.5
  • Length (cm)28
  • Width (cm)21.6
  • Thickness (mil)2.25
  • DescriptionAutoclave Biohazard Bags, Red, 8.5" x 11", PP; 100/PK
Reviews
Q&A