Cole-Parmer Autoclavable Biohazard Bags, Red, 12.2" x 26", PP; 200/CS

Representative image only.

Cole-Parmer Autoclavable Biohazard Bags, Red, 12.2" x 26", PP; 200/CS

Cole-Parmer | Item: 1400306

Heavy-gauge, 2.25 mil thick polypropylene (PP) bags offer excellent puncture resistance

$162.07CAD / Case of 200

$162.07CAD / Case of 200
Product Description

Key Features

  • Withstand temperatures up to 136°C (277°F)
  • Prominently marked with the words “Hazardous Waste” and the “Biohazard” symbol
  • White marking area for quick and easy labeling
  • 3 gallon capacity
  • Measures 12.2"W x 26"L (31 x 66 cm)
  • Case of 200 bags

More About this Item

These high-temperature autoclavable bags are extremely pliable yet offer exceptional strength and durability. Seamless bottoms ensure leak-free autoclaving. Opaque "biohazard" red color along with the words "Hazardous Waste" and the "biohazard" symbol conforms to OSHA standard (29CFR1910.1030). An oversized white marking area makes it easy to label each bag with a permanent marker.

Bags are sold 200/case with four inner packs of 50 bags for convenient dispensing and storage.

Specifications
  • ColorRed
  • MaterialPolypropylene
  • Capacity (Gal)3
  • Length (in)26
  • Width (in)12.2
  • Length (cm)66
  • Width (cm)31
  • Thickness (mil)2.25
  • DescriptionAutoclavable Biohazard Bags, Red, 12.2" x 26", PP; 200/CS
Reviews
Q&A