Cole-Parmer ATS20100 Reflux/Inerting Unit for Integrity 10
Representative image only.
Cole-Parmer ATS20100 Reflux/Inerting Unit for Integrity 10
Cole-Parmer | Item: 3663097
Keep your reaction station running smoothly with these parts and accessories
$19,507.68CAD / Each
$19,507.68CAD / Each
Product Description
Key Features
  • Suitable for use with Cole-Parmer® Integrity 10, RS-200, or RS-250 series reaction stations
  • Sample volume 1 to 25 mL
  • Temperature range of -30 to 150°C
  • 1 year warranty
  • Previous part number ATS20100
More About this Item
Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
The unit features 10 positions with a cell cavity diamter of 25.5 mm. Includes a stir speed range of 350 to 1200 rpm.
1 Year Warranty
Specifications
  • Compatible WithCole-Parmer Integrity 10 Reaction Station
  • DescriptionReflux/Inerting Unit for Integrity 10