Cole-Parmer Aluminum Cooling Block, 384-Well PCR Plate
Representative image only.
Cole-Parmer Aluminum Cooling Block, 384-Well PCR Plate
Cole-Parmer | Item: 6361505
Quit running to the freezer for more ice
$189.00CAD / Each
$300.94CAD / Each
$189.00CAD / Each
$300.94CAD / Each
Buy It Together
Current Item
Cole-Parmer Aluminum Cooling Block, 384-Well PCR Plate
Cole-Parmer Aluminum Cooling Block, 384-Well PCR Plate
6361505
$189.00CAD / Each
$300.94CAD / Each
+ +
Cole-Parmer Transport Box, 5 L
0372604
$559.15CAD / Each
+ =
Total Price 1214.48 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cool or heat your samples quickly—blocks adjust to desired temperature within minutes
  • Eliminate uneven cooling
  • Keep samples upright and organized while controlling temperature
  • Contoured wells hold tubes and vials snugly for excellent temperature convection
More About this Item
Place blocks on ice, dry ice, liquid nitrogen, or in a water bath. These anodized aluminum blocks can keep samples at or near 1°C for up to 8 hours in properly packed ice, or for 3 to 7 hours in dry ice. Choose the blocks that meet your application needs — configurations are available for 1.5-/2.0-mL microcentrifuge tubes and cryogenic vials, 0.5-mL tubes, and 96-well or 364-well plates.
Specifications
  • Tubes Held384
  • Tube SizePCR Plates
  • ColorBlue
  • DescriptionAluminum Cooling Block, 384-Well PCR Plate