Cole-Parmer 96-well DispoDialyzer, membrane MWCO 10,000 Daltons

Representative image only.

Cole-Parmer 96-well DispoDialyzer, membrane MWCO 10,000 Daltons

Cole-Parmer | Item: 0289286

Save time — simultaneously prepare 96 samples from 25µl to 300µl

Call For Price

Call For Price
Product Description

Key Features

  • Over 95% sample recovery
  • Each well contains a separate membrane to eliminate sample contamination and leakage
  • Very high reproducibility between different wells
  • 1 year warranty

More About this Item

These 96-well dialyzers let you simultaneously prepare 96 samples. Use for applications such as salt removal, buffer exchange, parallel sample prep after fraction collection, detergent removal, HPLC, and more.
Each well contains an individual, high-quality, regenerated cellulose membrane that eliminates sample contamination and leakage. Membranes are free from sulfur and heavy metal contamination. Wells are sealed with 8-cap strips (included).
To use the dialyzer, simply place the sample into each well and assemble the strip caps. Float the entire plate in dialysis buffer. Then, retrieve the purified samples for downstream applications.
What’s included: twelve 8-cap strips to seal wells.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)10000
  • Max Sample Volume (mL)0.3
  • Description96-well DispoDialyzer, membrane MWCO 10,000 Daltons
  • Warranty1 year
Reviews
Q&A