Cole-Parmer 2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 2 mL; 50/Pk

Representative image only.

Cole-Parmer 2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 2 mL; 50/Pk

Cole-Parmer | Item: 0439582

Embedded Datamatrix code provides fast and accurate sample processing

$213.64CAD / Pkg of 50

$213.64CAD / Pkg of 50

Buy It Together

Current Item

Cole-Parmer 2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 2 mL; 50/Pk
Cole-Parmer 2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 2 mL; 50/Pk
0439582

$213.64CAD / Pkg of 50
+ + =
Total Price 555.25 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Innovative “channels of evacuation” design allows fluid to easily drain from the vials

More About this Item

The cryogenic 2D vials, available in 2 and 5 mL sizes with internally or externally threaded caps, are idea for rapid processing of biological samples. The embedded Datamatrix code at the bottom of each vial along with the corresponding “Code 128” barcode on the body of the vial, saves time and provides accurate organization and tracking of valuable samples.

The 2D vials can be safely stored in polycarbonate boxes (purchase separately). The storage boxes also feature the 2D Datamatrix code and Code 128 barcode to provide automatic identification of both the box and stored vials.


Rated for use down to -196°C. Liquid phase storage may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. It is recommended storing in the gas phase above the nitrogen (-178 to –150 ºC) to prevent issue. Recommend filling the tube with no more than 90% of the total nominal volume to permit sample expansion.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications
  • Volume (mL)2
  • Tube MaterialPolypropylene
  • Tube Bottom ShapeSkirted
  • Cap TypeScrew cap
  • Cap MaterialPolyethylene and rubber
  • ThreadInternal
  • SterileYes
  • AutoclavableYes
  • Height (mm)47
  • Outside Diameter (mm)12.6
  • Description2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 2 mL; 50/Pk
Reviews
Q&A