Clift Industries 3500-035 Laboratory Safety Spill Kit; 3.5 Gal.; 1 Pack/case
Representative image only.
Clift Industries 3500-035 Laboratory Safety Spill Kit; 3.5 Gal.; 1 Pack/case
Clift Industries | Mfr: 3500-035| Item: 8629507
Contains everything needed to safely and easily remove hazardous spills
$442.75CAD / Each
$442.75CAD / Each
Product Description
Includes
1 Qt. Ultimate Acid Eater, 1 Qt. Base Eater, 1 Qt. HF Acid Eater, 1 Qt. BioRem-2000 Surface Cleaner, 1 Qt. Halex, Goggles, 8 mil vinyl aprons, 18 mil latex gloves, industrial sponge, sprayer applicator
Key Features
  • 2 year warranty
More About this Item
Laboratory Safety Spill Kit neutralizes organic and inorganic acids, hydrofluoric acid, caustics-bases, halogens and solvents, as well as vapors and odors. Neutralizing agent converts hazardous chemicals to safe, non-toxic organic salts that can be easily disposed of, eliminating disposal costs and safety issues. PH indicators eliminate need for pH meters or strips.
Two year warranty
Specifications
  • DescriptionLaboratory Safety Spill Kit; 3.5 Gal.; 1 Pack/case