Clift Industries 2900-025 HF Acid Eater Neutralizer, 2.5 Gallon; 2 Packs/Case
Representative image only.
Clift Industries 2900-025 HF Acid Eater Neutralizer, 2.5 Gallon; 2 Packs/Case
Clift Industries | Mfr: 2900-025| Item: 8629503
Neutralizes Hydrofluoric Acid into safe Organic salts
$767.64CAD / Case of 2
$767.64CAD / Case of 2
Product Description
Key Features
  • 2 year warranty
More About this Item
Unlike caustic soda and soda ash, which forms poisonous bi-fluoride, HF Acid Eater completely destroys the hydrofluoric acid polymer. This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda. pH indicator additive eliminates the need for additional pH strips taking out the guess work. HF Acid Eater is nonhazardous and noncorrosive, and is excellent for a variety of uses and applications. Apply on horizontal, vertical, and perforated surfaces. Also neutralizes vapors and reduces odors.
Two year warranty
Specifications
  • DescriptionHF Acid Eater Neutralizer Kit, 2.5 Gallon; 2 Packs/Case