Clift Industries 2002-005 Ultimate Acid Eater Spill Kit, 2 Gallon; 1/Cs
Representative image only.
Clift Industries 2002-005 Ultimate Acid Eater Spill Kit, 2 Gallon; 1/Cs
Clift Industries | Mfr: 2002-005| Item: 8629502
Eliminates the need for powders, sorbent pads, booms, and socks, as well as caustics
$497.27CAD / Each
$497.27CAD / Each
Product Description
Includes
Kit includes 2 gallons of Ultimate Acid Eater, 2 pair of hazmat boots, 2 pair of goggles, 2 pair of gloves, 2 aprons, MSDS, instructions
Key Features
  • 2 year warranty
More About this Item
Ultimate Acid Eater neutralizes hazardous acids in a quick, safe, and effective manner by transforming them into disposable organic salts. This material will also neutralize dangerous acid vapors that pose safety concerns. It has a 1:1 neutralization ratio or less with a variety of acids. pH color indicator additive enables you to monitor the neutralization process without pH instruments. Neutralizer is easy to apply on vertical surfaces, hard-to-reach areas, and permeable surfaces. Safe to use and store, this nonhazardous, noncorrosive, biodegradable, and environmentally safe product, dries quickly and will not leave any residues or stains. It also eliminates disposal issues and cost as well as safety hazards associated with acid spills.
Two year warranty
Specifications
  • DescriptionUltimate Acid Eater Spill Kit, 2 Gallon; 1/Cs