Clift Industries 2001-025 Ultimate Acid Eater; 2.5 Gal. Bottle; 2 Bottles/Case
Representative image only.
Clift Industries 2001-025 Ultimate Acid Eater; 2.5 Gal. Bottle; 2 Bottles/Case
Clift Industries | Mfr: 2001-025| Item: 8629501
Eliminates the need for powders, sorbent pads, booms, and socks, as well as caustics
$767.64CAD / Case of 2
$767.64CAD / Case of 2
Product Description
Key Features
  • 2 year warranty
More About this Item
Ultimate Acid Eater neutralizes hazardous acids in a quick, safe, and effective manner by transforming them into disposable organic salts. This material will also neutralize dangerous acid vapors that pose safety concerns. It has a 1:1 neutralization ratio or less with a variety of acids. pH color indicator additive enables you to monitor the neutralization process without pH instruments. Neutralizer is easy to apply on vertical surfaces, hard-to-reach areas, and permeable surfaces. Safe to use and store, this nonhazardous, noncorrosive, biodegradable, and environmentally safe product, dries quickly and will not leave any residues or stains. It also eliminates disposal issues and cost as well as safety hazards associated with acid spills.
Two year warranty
Specifications
  • DescriptionUltimate Acid Eater; 2.5 Gal.; 2 Packs/case