Chromaflex Standard Chromatography Column, 24 mL, 30 cm L x 1.0 cm ID

Representative image only.

Chromaflex Standard Chromatography Column, 24 mL, 30 cm L x 1.0 cm ID

Chromaflex | Mfr: 420830-3000| Item: 3469603

Unique o-ring design provides a solvent resistant, leak-free seal

$581.99CAD / Each

$581.99CAD / Each

Buy It Together

Current Item

Chromaflex Standard Chromatography Column, 24 mL, 30 cm L x 1.0 cm ID
Chromaflex Standard Chromatography Column, 24 mL, 30 cm L x 1.0 cm ID
3469603

$581.99CAD / Each
+ =
Total Price 604.53 ADD 2 ITEMS TO CART
Product Description

Includes

two PTFE end fittings with 20 µm porosity HDPE bed supports, screw caps, five feet (1.5 m) PTFE tubing, two flangeless tubing nuts and two flangeless ferrules.

Key Features

  • Constructed of borosilicate glass 3.3, PTFE, and HDPE for exceptional chemical resistance
  • Standard column features a glass barrel only, without a water jacket
  • Column has a 24 mL capacity

More About this Item

CHROMAFLEX® chromatography columns feature uniquely designed PTFE shielded o-rings to form a solvent resistant, leak-free seal. Each end fitting has a thin PTFE layer that is curled over the o-ring. Columns can be used with all aqueous buffers without modification and with organic solvents when the optional PTFE bed supports are installed in the end fittings and flow adapters. Columns also feature borosilicate glass barrel construction with PTFE end fittings and HDPE bed supports. Recommended sterilization is with ethylene oxide, 2N NaOH or 100% ethanol (autoclaving is not recommended).

Choose from standard or jacketed columns. Jacketed columns feature a water jacket around the barrel to provide thermal control of temperature-sensitive chromatographic separations. The jacket also acts as a safety shield.

Specifications
  • Column Length (cm)30
  • Column Length (mm)300
  • Column Inside Diameter (ID) (mm)10
  • Max Pressure (PSI)100
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)50
  • Total Volume Capacity (mL)24
  • MethodologyStandard Chromatography Column
  • DescriptionStandard Chromatography Column, 24 mL, 30 cm L x 1.0 cm ID
Reviews
Q&A