Chemetrics R8515 Refill Kit , Phosphate Test, for 00290-52
Representative image only.
Chemetrics R8515 Refill Kit , Phosphate Test, for 00290-52
Chemetrics | Mfr: R8515| Item: 0029053
Take it wherever you go
$73.67CAD / Box of 30
$73.67CAD / Box of 30
Buy It Together
Current Item
Chemetrics R8515 Refill Kit , Phosphate Test, for 00290-52
Chemetrics R8515 Refill Kit , Phosphate Test, for 00290-52
0029053
$73.67CAD / Box of 30
+ + +
Chemetrics K-9010 Colorimetric Silica Test Kit
0554104
$152.27CAD / Each
=
Total Price 444.29 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Specifications
  • Compatible With00290-52
  • DescriptionRefill Kit , Phosphate Test, for 00290-52