Chemetrics R-8510 Refill Kit , Phosphate Test, for 05540-92, Six-Month Shelf Life
Representative image only.
Chemetrics R-8510 Refill Kit , Phosphate Test, for 05540-92, Six-Month Shelf Life
Chemetrics | Mfr: R-8510| Item: 0554093
Take it wherever you go
$74.92CAD / Box of 1
$74.92CAD / Box of 1
Buy It Together
Current Item
Chemetrics R-8510 Refill Kit , Phosphate Test, for 05540-92, Six-Month Shelf Life
Chemetrics R-8510 Refill Kit , Phosphate Test, for 05540-92, Six-Month Shelf Life
0554093
$74.92CAD / Box of 1
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
$152.27CAD / Each
+ + =
Total Price 439.06 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
  • 2 year warranty
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Two year warranty
Specifications
  • Compatible With05540-92
  • DescriptionRefill Kit , Phosphate Test, for 05540-92, Six-Month Shelf Life