Chemetrics K-9510 Colorimetric Sufide Test Kit
Representative image only.
Chemetrics K-9510 Colorimetric Sufide Test Kit
Chemetrics | Mfr: K-9510| Item: 0554052
CHEMets test kits are portable and self-contained
$149.77CAD / Each
$149.77CAD / Each
Buy It Together
Current Item
Chemetrics K-9510 Colorimetric Sufide Test Kit
Chemetrics K-9510 Colorimetric Sufide Test Kit
0554052
$149.77CAD / Each
+ + + =
Total Price 394.42 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
More About this Item
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.
Specifications
  • TestSulfide, total soluble
  • Measurement Points0 to 1 ppm; 1 to 10 ppm
  • Tests Per Kit30
  • DescriptionColorimetric Sufide Test Kit