Chemetrics K-8510 Colorimetric Phosphate Test Kit

Representative image only.

Chemetrics K-8510 Colorimetric Phosphate Test Kit

Chemetrics | Mfr: K-8510| Item: 0554092

CHEMets test kits are portable and self-contained

$149.77CAD / Each

$149.77CAD / Each

Buy It Together

Current Item

Chemetrics K-8510 Colorimetric Phosphate Test Kit
Chemetrics K-8510 Colorimetric Phosphate Test Kit
0554092

$149.77CAD / Each
+ + +
LaMotte Insta-Test Alkalinity Test Strips, 0 to 180 ppm
9953262

$43.02CAD / Pkg of 50
=
Total Price 406.06 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Vacuum sealed reagents and handy comparator chart for quick, visual results
  • 2 year warranty

More About this Item

CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.

Two year warranty
Specifications
  • TestPhosphate
  • Measurement Points0 to 1 ppm; 1 to 10 ppm
  • Tests Per Kit30
  • DescriptionColorimetric Phosphate Test Kit
  • Warranty2 year
Documents & Certificates
Reviews
Q&A