Chemetrics K-2504 Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s

Representative image only.

Chemetrics K-2504 Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s

Chemetrics | Mfr: K-2504| Item: 0554090

CHEMets test kits are portable and self-contained

$152.27CAD / Each

$152.27CAD / Each

Buy It Together

Current Item

Chemetrics K-2504 Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s
Chemetrics K-2504 Test Kit Chlorine 0 to 1, 1 to 5 ppm Range s
0554090

$152.27CAD / Each
+ + + =
Total Price 396.92 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Vacuum sealed reagents and handy comparator chart for quick, visual results
  • 2 year warranty

More About this Item

CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.

Two year warranty
Specifications
  • TestChlorine, free and total
  • Measurement Points0 to 1 ppm; 1 to 5 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit30
  • DescriptionTest Kit Chlorine 0 to 1, 1 to 5 ppm Range s
  • Warranty2 year
Documents & Certificates
Reviews
Q&A