Celline CL1000 Cell Culture System, One 1000 mL Media Reservoir

Representative image only.

Celline CL1000 Cell Culture System, One 1000 mL Media Reservoir

Celline | Mfr: WCL1000-1| Item: 1305220

Ideal for monoclonal antibody and cellular protein production—viable in vitro alternative to using animals

$593.78CAD / Case of 1

$593.78CAD / Case of 1
Product Description

Key Features

  • Separate growth and media chambers
  • 15-mL cell cultivation chamber ideal for small-scale production of monoclonal antibodies and recombinant protein expression
  • Accommodates over 108 cells/mL
  • 18 month warranty

More About this Item

The cell culture systems feature separate growth and media chambers which provide continuous supply of nutrients and direct oxygenation. The systems also eliminate waste buildup for high cell densities.

Nonpyrogenic18 Month Warranty
Specifications
  • Capacity (L)1
  • AutoclavableNo
  • DescriptionCell Culture System, One 1000 mL Media Reservoir
  • Warranty18 months
Reviews
Q&A