Cell Spreader, Stainless Steel, 16-mm Spreading Bar

Representative image only.

Cell Spreader, Stainless Steel, 16-mm Spreading Bar

| Item: 1421200

Spread bacterial cultures evenly

$32.56CAD / Each

$32.56CAD / Each
Product Description

Key Features

  • Constructed of durable and reusable stainless steel
  • Apply cultures over an entire petri dish or plate with the triangular-shaped spreader
  • Autoclave or flame sterilize
  • Ideal for use with 60-mm plates

More About this Item

Stainless steel spreaders have a smooth triangular end with straight spreading bar to reduce the risk of damaging the agar. Both small and large sizes are 302 mm (8") long.

Specifications
  • MaterialStainless Steel
  • Width (in)5/8
  • Length (in)8
  • Width (mm)16
  • Length (mm)302
  • DescriptionCell Spreader, Stainless Steel, 16-mm Spreading Bar
Reviews
Q&A