Carver 2091-5 Press Test Cylinder with Pellet Molds; 2-1/4" ID

Representative image only.

Carver 2091-5 Press Test Cylinder with Pellet Molds; 2-1/4" ID

Carver | Mfr: 2091-5| Item: 5962082

$2,556.33CAD / Each

$2,556.33CAD / Each

Buy It Together

Current Item

Carver 2091-5 Press Test Cylinder with Pellet Molds; 2-1/4" ID
Carver 2091-5 Press Test Cylinder with Pellet Molds; 2-1/4" ID
5962082

$2,556.33CAD / Each
+ + =
Total Price 11047.92 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Test Cylinders with Pellet Molds can be used for molding, tableting, crushing, extruding, or sample preparation. Stainless steel (SS) construction. Cylinders include base plunger, U-shaped ejector, SS receiving pan (3/4''D x 5-1/2'' dia.), and filters. Use pan and filters to collect sample residue to determine percentages. Order replacement filters separately.

1 Year Warranty
Specifications
  • Inside Diameter (in)2 1/4
  • DescriptionPress Test Cylinder with Pellet Molds; 2-1/4" ID
  • Warranty1 Year
Reviews
Q&A