Burkle 5365-4000 Dipping Bottle with Glass Container, Nickel-Plated Brass Frame, 148 mm; 1000 mL

Representative image only.

Burkle 5365-4000 Dipping Bottle with Glass Container, Nickel-Plated Brass Frame, 148 mm; 1000 mL

Burkle | Mfr: 5365-4000| Item: 0604222

Ideal for point sampling of liquids

$1,480.76CAD / Each

$1,480.76CAD / Each

Buy It Together

Current Item

Burkle 5365-4000 Dipping Bottle with Glass Container, Nickel-Plated Brass Frame, 148 mm; 1000 mL
Burkle 5365-4000 Dipping Bottle with Glass Container, Nickel-Plated Brass Frame, 148 mm; 1000 mL
0604222

$1,480.76CAD / Each
+ =
Total Price 1741.18 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Suitable for Explosion Categories IIA and IIB
  • Tested for use in Ex protection zone 0
  • 15 month warranty

More About this Item

This dipping bottle with transparent glass container is ideal for sampling sewage, mud, and water. It can also test crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants of hazard categories IIA and IIB. The dipping frame is constructed of nickel-plated brass and features an easy-click closure made of electrically conductive polyamide.

15 Month Warranty
Specifications
  • MaterialNickel-plated brass frame
  • Sample Volume1000 mL
  • Length (in)13
  • Diameter (in)5.875
  • Length (cm)33
  • Diameter (cm)14.8
  • DescriptionDipping Bottle with Glass Container, Nickel-Plated Brass Frame, 148 mm; 1000 mL
  • Warranty15 month
Reviews
Q&A