BriskHeat THERMO-CUBE Thermostatically controlled power outlet, On at 35F / Off at 45F
Representative image only.
BriskHeat THERMO-CUBE Thermostatically controlled power outlet, On at 35F / Off at 45F
BriskHeat | Mfr: THERMO-CUBE| Item: 3611548
Ideal for freeze protection and thawing: metal or plastic pipes and valves
$32.12CAD / Each
$32.12CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Optimize the performance of your Briskheat instrumentation with these temperature protection accessories.
1 Year Warranty
Specifications
  • Max Temperature (° F)45
  • Max Temperature (° C)7
  • Min Temperature35;2
  • Plug TypeNo plug
  • Power (VAC)120
  • Power (Hz)50
  • DescriptionThermostatically controlled power outlet, On at 35F / Off at 45F