BriskHeat SRX12247201PT4 Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 12x24" , 120V, 720W

Representative image only.

BriskHeat SRX12247201PT4 Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 12x24" , 120V, 720W

BriskHeat | Mfr: SRX12247201PT4A| Item: 3600344

FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G

$1,007.80CAD / Each

$1,007.80CAD / Each
Product Description

Key Features

  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
  • easy-to-apply industrial strength pressure sensitive adhesive backing
  • 1 year warranty

More About this Item

These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces.

Approvals and compliances: FM, RoHS, REACH

Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.

1 Year Warranty
Specifications
  • Max Temperature (° F)158
  • Max Temperature (° C)70
  • Plug TypeNo plug
  • Power (VAC)120
  • Power (Hz)50-60
  • Power (amps)6
  • Power (watts)720
  • Width (in)12
  • Length (in)24
  • Lead Length (in)12
  • DescriptionHazardous T4A-Rated Silicone Heating Blanket, Adhesive, 12x24" , 120V, 720W
  • Warranty1 year
Reviews
Q&A