BriskHeat FGDHC55120 55 gallon, Full Coverage, Drum Heater - 120V

Representative image only.

BriskHeat FGDHC55120 55 gallon, Full Coverage, Drum Heater - 120V

BriskHeat | Mfr: FGDHC55120D| Item: 0930050

Provides safe, uniform heat to 55-gallon metal or plastic drums

$1,628.10CAD / Each

$1,628.10CAD / Each

Buy It Together

Current Item

BriskHeat FGDHC55120 55 gallon, Full Coverage, Drum Heater - 120V
BriskHeat FGDHC55120 55 gallon, Full Coverage, Drum Heater - 120V
0930050

$1,628.10CAD / Each
+ =
Total Price 1793.86 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Built-in adjustable thermoswitch prevents temperature runaway Lightweight flexible design makes installation quick and easy Order Insulation Cover (09300-70) separately
  • 1 year warranty

More About this Item

These insulated drum heaters provide a safe and efficient method of temperature and viscosity control for liquids in 55-gallon drums. Simply wrap insulated drum heater around a 55-gallon drum, adjust the built-in thermostat to desired temperature setting, and uniform heat is evenly distributed around the entire drum. The grounded heating element provides a high-watt density, ensuring rapid heating times — one inch thick fiberglass insulation prevents heat loss. Prevent further heat loss by ordering the drum insulation top cover (optional) below.
Drum heaters feature a hook-and-loop seal for easy installation and removal and models are available for both metal and plastic 55-gallon drums. Heaters come with standard three-prong plug (240 VAC models do not come with plugs) and are intended for indoor use in non-hazardous areas only.

Not CSA approved

1 Year Warranty
Specifications
  • Drum MaterialMetal
  • Drum Size55 gallons
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Min Temperature50;10
  • Plug TypeUS Standard
  • Power (VAC)120
  • Power (amps)13.3
  • Power (watts)1600
  • Diameter (in)22.3 in
  • Description55gallon, Full Coverage, Drum Heater - 120V
  • Warranty1 year
Reviews
Q&A