BriskHeat CPHTC1030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 120 V

Representative image only.

BriskHeat CPHTC1030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 120 V

BriskHeat | Mfr: CPHTC1030| Item: 0312230

Spiral wound heating element-packs more heating element into each inch of cord.

$291.36CAD / Each

$291.36CAD / Each
Product Description

Key Features

  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
  • 1 year warranty

More About this Item

Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications.

A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord. Cords feature low power consumption: the 3-,6-, and 12-ft 120 VAC cords (heating 1/4", 1/2", and 1" glass tubing respectively) draw about 0.1 watt/inch at full voltage/high temperature.

Control heating cord temperature with our DIGI-SENSE® temperature controller model 89800-01 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads.

Effective Oct-2014 these heating cords will come with a 120 VAC split plug that came be pluged into a standard oulet. Cords produced before Oct-2014 have a tip plugs and require the power cord adapter 03122-71

1 Year Warranty
Specifications
  • Max Temperature (° F)850
  • Max Temperature (° C)454
  • Plug TypeSplit plug
  • Power (VAC)120
  • Power (watts)53
  • Width (in)3/16
  • Length (ft)3
  • DescriptionFlexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 120 V
  • Warranty1 year
Reviews
Q&A