BriskHeat CPBIH102060LGT Grounded Fiberglass Insulated Heat Tape, 6' x 1" , 300 Watts, 230 V

Representative image only.

BriskHeat CPBIH102060LGT Grounded Fiberglass Insulated Heat Tape, 6' x 1" , 300 Watts, 230 V

BriskHeat | Mfr: CPBIH102060LGT| Item: 0312142

Safe for heating metal vessels

$235.37CAD / Each

$235.37CAD / Each
Product Description

Key Features

  • Heat to 482F (250C)
  • NOTE: Temperature range has been reduced to meet safty standards
  • 1 year warranty

More About this Item

Wind heating tape around glass, ceramic, or metal vessels—grounding prevents shock hazard. Temperature-resistant fiberglass sleeves encase heating element; ground wire at core. The maximum surface loading is 4 watts/sq in. Leads are 31/2" long with electrical connector at one end. Tapes include ties at both ends.

WARNING: Heating tape should not be overlapped and require the use of a temperature or power controller sold separately.

1 Year Warranty
Specifications
  • Max Temperature (° F)482
  • Max Temperature (° C)250
  • Plug TypeNo plug
  • Power (VAC)230
  • Power (watts)300
  • Width (in)1
  • Length (ft)6
  • DescriptionGrounded Fiberglass Insulated Heat Tape, 6' x 1" , 300 Watts, 230 V
  • Warranty1 year
Reviews
Q&A