Branson CPN-955-007 General Purpose Powder Cleaning Solution, 2 lb. container
Representative image only.
Branson CPN-955-007 General Purpose Powder Cleaning Solution, 2 lb. container
Branson | Mfr: CPN-955-007| Item: 0880202
Two pounds of concentrate makes 75 gallons of solution
$112.71CAD / Each
$112.71CAD / Each
Product Description
More About this Item
Non-corrosive powder concentrate is biodegradable and caustic-free. Removes dust, oils, light greases, and soils from aluminum, plastic, rubber, glass and more. Dilute with three ounces of water to generate one gallon of solution; Scoop included.
Specifications
  • FormatPowder
  • DescriptionGeneral Purpose Powder Cleaning Solution, 2 lb. container