BrandTech Transferpette® Electronic Pipette AC Adapter, US and Japan; 110 VAC, 50/60 Hz

Representative image only.

BrandTech Transferpette® Electronic Pipette AC Adapter, US and Japan; 110 VAC, 50/60 Hz

BrandTech | Mfr: 705352| Item: 2501660

Optimize performance of your electronic pipettors

$209.59CAD / Each

$209.59CAD / Each

Buy It Together

Current Item

BrandTech Transferpette® Electronic Pipette AC Adapter, US and Japan; 110 VAC, 50/60 Hz
BrandTech Transferpette® Electronic Pipette AC Adapter, US and Japan; 110 VAC, 50/60 Hz
2501660

$209.59CAD / Each
+ =
Total Price 1264.54 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Compatible with the BrandTech® Transferpette® pipettes

More About this Item

Choose from a wide range of accessories and replacement parts to extend the life and enhance operation of your Transferpette® electronic pipettors.

Specifications
  • Compatible WithTransferpette® Electronic Pipette
  • ConnectionsUS and Japan
  • Power (VAC)110
  • Power (Hz)50/60
  • DescriptionTransferpette® Electronic Pipette AC Adapter, US and Japan; 110 VAC, 50/60 Hz
Reviews
Q&A