BrandTech Seripettor Economy Dispenser, 2.5 to 25 mL

Representative image only.

BrandTech Seripettor Economy Dispenser, 2.5 to 25 mL

BrandTech | Mfr: 4720150| Item: 0787832

An economical choice for dispensing of many liquids in biological labs

$359.88CAD / Each

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$359.88CAD / Each

Buy It Together

Current Item

BrandTech Seripettor Economy Dispenser, 2.5 to 25 mL
BrandTech Seripettor Economy Dispenser, 2.5 to 25 mL
0787832

$359.88CAD / Each
+ + =
Total Price 700.60 ADD 3 ITEMS TO CART
Product Description

Includes

Dispenser with 45mm thread, thread adapters to fit S40, 38 mm and 33 mm threads, replacement piston/cylinder cartridge, 250 mm filling tube, and manual.

Key Features

  • Spring-loaded piston for one handed dispensing
  • Replaceable piston/cylinder cartridge to renew dispenser
  • Suitable for sodium azide solutions
  • 1 year warranty

More About this Item

The seripettor® is an economical dispenser suitable for many needs of the biology laboratory. An excellent choice for bases, buffers, and other aqueous solutions as well as dilute, non-oxidizing acids, and polar solvents. Wetted materials include PP, HDPE, and EPDM rubber. Not recommended for concentrated acids, non-polar solvents, or scintillation fluid. Accessories are available for dispensing of culture media or agar.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year WarrantyISO9001 Certified Supplier
Specifications
  • Min Dispense Volume (mL)2.5
  • Max Dispense Volume (mL)25
  • Division (mL)0.5
  • Accuracy±1.2%
  • Reproducibility±0.2%
  • AutoclavableLower body only
  • Wetted PartsPP, PE, and EPDM
  • DescriptionSeripettor Economy Dispenser, 2.5 to 25 mL
  • Warranty1 year
Reviews
Q&A