BrandTech 4720450 Seripettor® Pro Dispenser, 2.5 to 25 mL

Representative image only.

BrandTech 4720450 Seripettor® Pro Dispenser, 2.5 to 25 mL

BrandTech | Mfr: 4720450| Item: 0787833

Wider range of liquids than standard seripettor model

$1,004.85CAD / Each

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,004.85CAD / Each

Buy It Together

Current Item

BrandTech 4720450 Seripettor® Pro Dispenser, 2.5 to 25 mL
BrandTech 4720450 Seripettor® Pro Dispenser, 2.5 to 25 mL
0787833

$1,004.85CAD / Each
+ + =
Total Price 1289.80 ADD 3 ITEMS TO CART
Product Description

Includes

Spare dispensing cartridge, discharge tube, telescoping filling tube, and three polypropylene adapters (S30, 33 & 38 mm).

Key Features

  • Replaceable dispensing cartridge keeps service costs down
  • Opaque upper sleeve protects light-sensitive reagents
  • 1 year warranty

More About this Item

These seripettor® pro dispensers offer the rugged versatility and low lifetime cost of the regular seripettor and handle a wider range of liquids. Ideal for customers who do not require all the features of the Dispensette® dispensers. Seripettor pro dispensers are threaded to fit directly onto 45-mm screw-neck bottles; adapters for S40, 33- and 38-mm bottles are included. Order bottles separately.

1 Year WarrantyISO9001 Certified Supplier
Specifications
  • Min Dispense Volume (mL)2.5
  • Max Dispense Volume (mL)25
  • Division (mL)0.5
  • Accuracy±1.2%
  • Reproducibility±0.2%
  • AutoclavableLower body only
  • Wetted PartsPP, PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium and borosilicate glass
  • Descriptionseripettor pro Dispenser, 2.5 to 25 mL
  • Warranty1 year
Reviews
Q&A