Bel-Art Poxygrid Benchtop Biohazard Bag Holder Kit

Representative image only.

Bel-Art Poxygrid Benchtop Biohazard Bag Holder Kit

Bel-Art | Mfr: F13193-0500| Item: 0649601

Comply with OSHA standards for biohazardous material handling

$111.39CAD / Each

$111.39CAD / Each
Product Description

Includes

Cover and one hundred 8-1/2" x 11" (21.6 x 27.9 cm) biohazard bags.

Key Features

  • Epoxy-coated steel wire stand is durable and easy to clean
  • All components are autoclavable
  • Snug-fitting cover secures bag to the stand--does not impede safe bag removal
  • 3 month warranty

More About this Item

These orange colored epoxy-coated steel wire stands can securely hold a high-density 8 1/2" x 11" (22 x 28 cm) polyethylene bag in an upright open position. Ideal for collecting pipette tips, microcentrifuge tubes, and other hazardous materials throughout the day. The stands are reusable, can be cleaned by hand or in automated dish washer, and autoclavable at 250°F (121°C).

The biohazard bag holder kit features a snap-on polypropylene cover which fits snugly enough to provide stable containment, but is easy enough to disassemble and won't lead to spills or torn bags. Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes.

3 Month Warranty
Specifications
  • MaterialEpoxy-coated steel wire and polypropylene cover
  • ColorOrange
  • Length (in)11
  • Width (in)8.5
  • Length (cm)28
  • Width (cm)22
  • Thickness (mil)0.71
  • DescriptionPoxygrid Benchtop Biohazard Bag Stand with Cover
  • Warranty3 month
Reviews
Q&A